Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IHH
Entrez Gene ID
488533
Gene Name
Indian hedgehog signaling molecule
Gene Aliases
-
Chromosome Location
Chr.37: 25616942 - 25622989 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_545653.6 XP_545653.3 3 - 3 717 137

View Full Product Details