Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNS3
Entrez Gene ID
482982
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Gene Aliases
-
Chromosome Location
Chr.17: 13297242 - 13338319 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630152.3 XP_005630209.1 3 - 3 816 135
XM_005630154.3 XP_005630211.1 3 - 3 807 135
XM_014120544.2 XP_013976019.1 3 - 3 766 135
XM_022404473.1 XP_022260181.1 2 - 2 631 135
XM_540095.6 XP_540095.1 3 - 3 816 135

View Full Product Details