Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRX5
Entrez Gene ID
487276
Gene Name
iroquois homeobox 5
Gene Aliases
-
Chromosome Location
Chr.2: 60957353 - 60978782 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617547.3 XP_005617604.1 2 - 3 1158 134
XM_022411225.1 XP_022266933.1 2 - 3 1158 134
XM_846105.4 XP_851198.1 2 - 3 1157 134

View Full Product Details