Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAME
Entrez Gene ID
477552
Gene Name
PRAME nuclear receptor transcriptional regulator
Gene Aliases
-
Chromosome Location
Chr.26: 27124901 - 27137772 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636545.2 XP_005636602.1 3 - 4 698 189
XM_005636546.2 XP_005636603.1 3 - 4 481 189
XM_014108142.2 XP_013963617.1 3 - 4 407 189
XM_022410565.1 XP_022266273.1 4 - 5 599 189
XM_022410566.1 XP_022266274.1 4 - 5 626 189
XM_022410567.1 XP_022266275.1 2 - 3 635 189

View Full Product Details