Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL3
Entrez Gene ID
610736
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.15: 16324752 - 16434256 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022428053.1 XP_022283761.1 9 - 10 898 79

View Full Product Details