Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP24
Entrez Gene ID
485849
Gene Name
matrix metallopeptidase 24
Gene Aliases
-
Chromosome Location
Chr.24: 24223049 - 24258082 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634968.3 XP_005635025.2 4 - 5 450 75

View Full Product Details