Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBX3
Entrez Gene ID
482015
Gene Name
chromobox 3
Gene Aliases
-
Chromosome Location
Chr.14: 39510694 - 39520796 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628649.3 XP_005628706.1 2 - 3 134 184
XM_022427500.1 XP_022283208.1 1 - 2 641 184
XM_859382.5 XP_864475.3 1 - 2 636 184

View Full Product Details