Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RYBP
Entrez Gene ID
484690
Gene Name
RING1 and YY1 binding protein
Gene Aliases
-
Chromosome Location
Chr.20: 19650936 - 19732987 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407339.1 XP_022263047.1 - 756 117

View Full Product Details