Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR3
Entrez Gene ID
490062
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.6: 38941933 - 38944110 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022420259.1 XP_022275967.1 2 - 3 435 72

View Full Product Details