Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEOX1
Entrez Gene ID
607271
Gene Name
mesenchyme homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 19614568 - 19641426 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624420.3 XP_005624477.1 - 208 151
XM_022422945.1 XP_022278653.1 - 555 151
XM_022422946.1 XP_022278654.1 - 599 151

View Full Product Details