Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DLX3
Entrez Gene ID
491073
Gene Name
distal-less homeobox 3
Gene Aliases
-
Chromosome Location
Chr.9: 26018101 - 26022964 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548193.5 XP_548193.2 - 477 137

View Full Product Details