Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF6
Entrez Gene ID
477332
Gene Name
ring finger protein 6
Gene Aliases
-
Chromosome Location
Chr.25: 13199201 - 13240347 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003433296.4 XP_003433344.1 - 1087 126
XM_005635533.3 XP_005635590.1 - 1104 126
XM_005635534.3 XP_005635591.1 - 1104 126
XM_005635535.3 XP_005635592.1 - 1187 126
XM_022409627.1 XP_022265335.1 - 1083 126
XM_022409628.1 XP_022265336.1 - 1100 126
XM_022409629.1 XP_022265337.1 - 1100 126
XM_534526.6 XP_534526.2 - 1191 126

View Full Product Details