Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLP1
Entrez Gene ID
610161
Gene Name
cleavage factor polyribonucleotide kinase subunit 1
Gene Aliases
-
Chromosome Location
Chr.18: 38528700 - 38531899 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631200.3 XP_005631257.1 2 - 3 982 144
XM_005631201.3 XP_005631258.1 2 - 3 934 144
XM_847579.5 XP_852672.1 2 - 3 785 144

View Full Product Details