Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
IL17C
Entrez Gene ID
608988
Gene Name
interleukin 17C
Gene Aliases
-
Chromosome Location
Chr.5: 64702668 - 64703694 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846163.3 XP_851256.1 - 323 182

View Full Product Details