Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VGLL4
Entrez Gene ID
607588
Gene Name
vestigial like family member 4
Gene Aliases
-
Chromosome Location
Chr.20: 6751951 - 6915650 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632147.3 XP_005632204.1 - 1430 144
XM_005632148.3 XP_005632205.1 - 1112 144
XM_005632150.3 XP_005632207.1 - 601 144
XM_014121565.2 XP_013977040.1 - 782 144
XM_022406363.1 XP_022262071.1 - 811 144
XM_844730.5 XP_849823.2 - 785 144

View Full Product Details