Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSKU
Entrez Gene ID
485178
Gene Name
tsukushi, small leucine rich proteoglycan
Gene Aliases
-
Chromosome Location
Chr.21: 21910333 - 21927758 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005633465.3 XP_005633522.3 2 - 2 643 101
XM_005633467.3 XP_005633524.1 2 - 2 4894 101
XM_022407657.1 XP_022263365.1 2 - 2 1622 101

View Full Product Details