Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FBXO40
Entrez Gene ID
488004
Gene Name
F-box protein 40
Gene Aliases
-
Chromosome Location
Chr.33: 24841998 - 24899973 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639548.3 XP_005639605.1 4 - 4 2467 156
XM_022414009.1 XP_022269717.1 5 - 5 2150 156
XM_022414010.1 XP_022269718.1 4 - 4 2201 156

View Full Product Details