Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLTP
Entrez Gene ID
100856786
Gene Name
glycolipid transfer protein
Gene Aliases
-
Chromosome Location
Chr.26: 17262008 - 17283735 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639917.4 XP_003639965.1 4 - 5 534 88

View Full Product Details