Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
482086
Gene Name
exosome component 4
Gene Aliases
-
Chromosome Location
Chr.13: 37574359 - 37576331 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539207.6 XP_539207.1 1 - 1 331 113

View Full Product Details