Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLX3
Entrez Gene ID
489123
Gene Name
T cell leukemia homeobox 3
Gene Aliases
-
Chromosome Location
Chr.4: 40816351 - 40818582 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546241.6 XP_546241.5 3 - 3 754 106

View Full Product Details