Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAND2
Entrez Gene ID
607431
Gene Name
heart and neural crest derivatives expressed 2
Gene Aliases
DHAND
Chromosome Location
Chr.25: 24140496 - 24143143 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410028.1 XP_022265736.1 1 - 2 434 147

View Full Product Details