Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPPR2
Entrez Gene ID
484944
Gene Name
phospholipid phosphatase related 2
Gene Aliases
LPPR2
Chromosome Location
Chr.20: 49955538 - 49964207 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632856.3 XP_005632913.1 6 - 7 1333 94
XM_022407023.1 XP_022262731.1 6 - 7 1333 94
XM_022407024.1 XP_022262732.1 6 - 7 1285 94
XM_022407025.1 XP_022262733.1 6 - 7 1333 94
XM_022407026.1 XP_022262734.1 6 - 7 1387 94
XM_022407027.1 XP_022262735.1 5 - 6 1212 94
XM_022407028.1 XP_022262736.1 6 - 7 1427 94
XM_022407029.1 XP_022262737.1 6 - 7 1333 94
XM_022407030.1 XP_022262738.1 6 - 7 1333 94

View Full Product Details