Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP1
Entrez Gene ID
613003
Gene Name
receptor accessory protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 38807217 - 38908434 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014120496.2 XP_013975971.1 2 - 3 219 95
XM_014120497.2 XP_013975972.1 2 - 3 201 95
XM_022404741.1 XP_022260449.1 2 - 3 218 95

View Full Product Details