Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HOXB9
- Entrez Gene ID
-
608958
- Gene Name
- homeobox B9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 24884101 - 24889904 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_859652.5 |
XP_864745.2 |
- |
987 |
98 |
- Gene Symbol
- HOXB8
- Entrez Gene ID
-
491059
- Gene Name
- homeobox B8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 24874893 - 24888981 on Build CanFam3.1
View Full Product Details