Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB8
Entrez Gene ID
486587
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
-
Chromosome Location
Chr.27: 6586933 - 6619240 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636909.3 XP_005636966.1 2 - 3 283 125
XM_005636910.3 XP_005636967.1 3 - 4 430 125
XM_005636911.3 XP_005636968.1 3 - 4 426 125
XM_005636913.3 XP_005636970.1 2 - 3 250 125
XM_022410952.1 XP_022266660.1 3 - 4 468 125
XM_022410953.1 XP_022266661.1 2 - 3 337 125
XM_543713.5 XP_543713.2 2 - 3 251 125
XM_858305.5 XP_863398.1 3 - 4 352 125

View Full Product Details