Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHF1
Entrez Gene ID
484612
Gene Name
pleckstrin homology and FYVE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 121776122 - 121802881 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616798.2 XP_005616855.1 4 - 4 673 157
XM_005616799.3 XP_005616856.1 4 - 4 3361 157
XM_005616800.3 XP_005616857.1 3 - 3 642 157
XM_005616802.3 XP_005616859.1 2 - 2 369 157
XM_014120944.2 XP_013976419.1 3 - 3 618 157
XM_014120950.2 XP_013976425.1 3 - 3 3290 157
XM_022426532.1 XP_022282240.1 5 - 5 3718 157
XM_022426536.1 XP_022282244.1 3 - 3 464 157
XM_022426537.1 XP_022282245.1 3 - 3 481 157
XM_541726.5 XP_541726.2 3 - 3 447 157

View Full Product Details