Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LENG1
Entrez Gene ID
476388
Gene Name
leukocyte receptor cluster member 1
Gene Aliases
-
Chromosome Location
Chr.1: 103054250 - 103057440 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533591.6 XP_533591.3 2 - 3 486 99

View Full Product Details