Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
482269
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 8738379 - 8743623 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539388.6 XP_539388.2 1 - 2 1432 168

View Full Product Details