Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC8C
Entrez Gene ID
490168
Gene Name
leucine rich repeat containing 8 VRAC subunit C
Gene Aliases
-
Chromosome Location
Chr.6: 58895769 - 58932729 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621951.3 XP_005622008.1 2 - 2 1812 137
XM_005621952.3 XP_005622009.1 3 - 3 1951 137
XM_547289.6 XP_547289.2 2 - 2 1745 137

View Full Product Details