Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIEN1
Entrez Gene ID
480532
Gene Name
migration and invasion enhancer 1
Gene Aliases
-
Chromosome Location
Chr.9: 22758493 - 22759935 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624552.2 XP_005624609.1 - 340 125

View Full Product Details