Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
WNT3A
Entrez Gene ID
482208
Gene Name
Wnt family member 3A
Gene Aliases
-
Chromosome Location
Chr.14: 864393 - 899698 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539327.5 XP_539327.3 7 - 7 970 153

View Full Product Details