Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELFN2
Entrez Gene ID
481269
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62
Chromosome Location
Chr.10: 27185104 - 27191461 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022424469.1 XP_022280177.1 1 - 1 2332 147

View Full Product Details