Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TENT5B
Entrez Gene ID
487348
Gene Name
terminal nucleotidyltransferase 5B
Gene Aliases
FAM46B
Chromosome Location
Chr.2: 73156894 - 73165745 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544474.6 XP_544474.3 2 - 2 1822 117

View Full Product Details