Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOSC6
Entrez Gene ID
489713
Gene Name
exosome component 6
Gene Aliases
-
Chromosome Location
Chr.5: 76286286 - 76287667 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620702.3 XP_005620759.1 1 - 1 217 147

View Full Product Details