Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF12
Entrez Gene ID
474992
Gene Name
serine and arginine rich splicing factor 12
Gene Aliases
SFRS13B
Chromosome Location
Chr.12: 48355844 - 48398753 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005627622.3 XP_005627679.1 5 - 5 954 122
XM_014118142.2 XP_013973617.1 5 - 5 635 122
XM_022426347.1 XP_022282055.1 5 - 5 793 122
XM_022426348.1 XP_022282056.1 5 - 5 600 122

View Full Product Details