Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXNL1
Entrez Gene ID
484836
Gene Name
nucleoredoxin like 1
Gene Aliases
TXNL6
Chromosome Location
Chr.20: 45348890 - 45351603 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541952.2 XP_541952.2 1 - 1 106 161

View Full Product Details