Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B3GNT6
Entrez Gene ID
485176
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3GNT1
Chromosome Location
Chr.21: 21693069 - 21694140 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407654.1 XP_022263362.1 1 - 1 744 168

View Full Product Details