Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDCD2
Entrez Gene ID
479302
Gene Name
NudC domain containing 2
Gene Aliases
-
Chromosome Location
Chr.4: 47787146 - 47793325 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536440.5 XP_536440.4 1 - 2 515 175

View Full Product Details