Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL14
Entrez Gene ID
486755
Gene Name
F-box and leucine rich repeat protein 14
Gene Aliases
-
Chromosome Location
Chr.27: 43617087 - 43625269 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637363.3 XP_005637420.1 1 - 1 1270 104
XM_022411125.1 XP_022266833.1 1 - 1 1270 104
XM_022411126.1 XP_022266834.1 1 - 1 1270 104
XM_022411128.1 XP_022266836.1 2 - 2 1079 104
XM_543882.6 XP_543882.2 1 - 1 1270 104

View Full Product Details