Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC16A14
Entrez Gene ID
486154
Gene Name
solute carrier family 16 member 14
Gene Aliases
-
Chromosome Location
Chr.25: 42316294 - 42373862 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022409647.1 XP_022265355.1 6 - 6 1734 85
XM_543280.5 XP_543280.2 4 - 4 548 85

View Full Product Details