Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSHZ2
Entrez Gene ID
106557707
Gene Name
teashirt zinc finger homeobox 2
Gene Aliases
-
Chromosome Location
Chr.24: 39124993 - 39354202 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022409042.1 XP_022264750.1 3 - 3 3414 100

View Full Product Details