Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTN4RL1
Entrez Gene ID
491197
Gene Name
reticulon 4 receptor like 1
Gene Aliases
-
Chromosome Location
Chr.9: 46061619 - 46133970 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624940.3 XP_005624997.1 2 - 2 1057 150

View Full Product Details