Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LANCL3
Entrez Gene ID
491823
Gene Name
LanC like 3
Gene Aliases
-
Chromosome Location
Chr.X: 32349007 - 32452130 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548943.6 XP_548943.2 1 - 2 832 137

View Full Product Details