Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB1
Entrez Gene ID
490731
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 38933721 - 38959306 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123330.1 NP_001116802.1 2 - 3 1988 100
GenBank mRNA EU541625.1 - 2 - 3 1988 100

View Full Product Details