Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2W1
Entrez Gene ID
491601
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
Chromosome Location
Chr.6: 15883248 - 15889508 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022419812.1 XP_022275520.1 - 970 89
XM_548722.5 XP_548722.2 - 968 89

View Full Product Details