Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKR1E2
Entrez Gene ID
608809
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2
Chromosome Location
Chr.2: 30694119 - 30706693 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617180.3 XP_005617237.1 - 466 92

View Full Product Details