Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESPN
Entrez Gene ID
489631
Gene Name
espin
Gene Aliases
-
Chromosome Location
Chr.5: 60285185 - 60316261 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620456.2 XP_005620513.1 - 428 93
XM_005620457.2 XP_005620514.1 - 428 93
XM_546751.5 XP_546751.2 - 301 93

View Full Product Details