Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNYL1
Entrez Gene ID
488492
Gene Name
cyclin Y like 1
Gene Aliases
-
Chromosome Location
Chr.37: 16075459 - 16113583 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005640591.3 XP_005640648.1 5 - 6 525 146

View Full Product Details