Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFAP2E
Entrez Gene ID
482482
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
-
Chromosome Location
Chr.15: 6552255 - 6556863 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119246.2 XP_013974721.1 4 - 5 513 180

View Full Product Details