Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN2
Entrez Gene ID
488749
Gene Name
zinc finger and SCAN domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 53864823 - 53905236 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014112447.2 XP_013967922.1 2 - 2 397 107
XM_022416955.1 XP_022272663.1 2 - 2 444 107
XM_545867.7 XP_545867.5 2 - 2 395 107

View Full Product Details